Tlačiť

Anti-CD47 Rabbit Recombinant Antibody [clone: DM212]

Dodávateľ: ANTIBODIES.COM
A318497-100 A318497-500
ANTIA318497-100EA 731.65 EUR
ANTIA318497-100 ANTIA318497-500
Anti-CD47 Rabbit Recombinant Antibody [clone: DM212]
Protilátky
Recombinant rabbit monoclonal [DM212] antibody to CD47 for ELISA and Flow Cytometry with samples derived from human.

  • Validated Applications: ELISA, Flow Cytometry
  • Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100

Type: Primary
Antigen: CD47
Clonality: Recombinant
Clone: DM212
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR