Tlačiť

Anti-IgG Antibody

Dodávateľ: ANTIBODIES.COM
A190-100 A190-200
ANTIA190-100EA 508.31 EUR
ANTIA190-100 ANTIA190-200
Anti-IgG Antibody
Protilátky
  • Validated applications: ELISA, WB
  • Recommended dilutions: ELISA: 1/1000 to 1/5000, WB: 1/500 to 1/10000

Type: Primary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR