Tlačiť

Anti-B7-H6 Rabbit Recombinant Antibody [clone: DM173]

Dodávateľ: ANTIBODIES.COM
A318536-100 A318536-500
ANTIA318536-100EA 731.65 EUR
ANTIA318536-100 ANTIA318536-500
Anti-B7-H6 Rabbit Recombinant Antibody [clone: DM173]
Protilátky
Recombinant rabbit monoclonal [DM173] antibody to B7-H6 for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: B7-H6
Clonality: Recombinant
Clone: DM173
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR