Tlačiť
ESD Safety Shoes, Lace-up, Low, X-Light, S3
Dodávateľ: Abeba
7131860/35 7131860/36 7131860/37 7131860/38 7131860/39 7131860/40 7131860/41 7131860/42 7131860/43 7131860/44 7131860/45 7131860/46 7131860/47 7131860/48
ABEB7131860/35EA 117.53 EUR
ABEB7131860/35 ABEB7131860/36 ABEB7131860/37 ABEB7131860/38 ABEB7131860/39 ABEB7131860/40 ABEB7131860/41 ABEB7131860/42 ABEB7131860/43 ABEB7131860/44 ABEB7131860/45 ABEB7131860/46 ABEB7131860/47 ABEB7131860/48
ESD Safety Shoes, Lace-up, Low, X-Light, S3
Safety shoes x-light is made up of microfibre, breathable inner lining with silver ions.
  • Reflective stripes
  • Instep area with lacing
  • Sole elevation possible
  • Metal-free penetration safety
  • X-light B ESD outsole black
  • Steel cap
  • Slip-resistant PU outsole
  • Breathable
ESD-compatible according to DIN EN 61340.
Certifikácie: EN ISO 20345:2011.