Tlačiť
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 540)
Dodávateľ: AAT BIOQUEST
10033120 10033121
AATB10033120EA 397.11 EUR
AATB10033120 AATB10033121
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 540)
Protilátky
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human