Tlačiť
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet 500)
Dodávateľ: AAT BIOQUEST
10441100 10441101
AATB10441100EA 397.11 EUR
AATB10441100 AATB10441101
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet 500)
Protilátky
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Affinity purified
  • Easily reconstituted
Pozor: For research use only.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ Violet 500
Epitope:
Host: Rat
Isotype:
Reactivity: Human