Tlačiť

Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 500)

Dodávateľ: AAT BIOQUEST
10141100 10141101
AATB10141100EA 397.11 EUR
AATB10141100 AATB10141101
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (mFluor™ Violet 500)
Protilátky
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Affinity purified
  • Easily reconstituted

Pozor: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR