Tlačiť
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Violet 540)
Dodávateľ: AAT Bioquest
10051120 10051121
AATB10051120EA 397.11 EUR
AATB10051120 AATB10051121
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Violet 540)
Protilátky
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human