Tlačiť
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Violet 510)
Dodávateľ: AAT BIOQUEST
10380110 10380111
AATB10380110EA 397.11 EUR
AATB10380110 AATB10380111
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Violet 510)
Protilátky
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human