Tlačiť
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 840)
Dodávateľ: AAT BIOQUEST
100410Q0 100410Q1
AATB100410Q0EA 397.11 EUR
AATB100410Q0 AATB100410Q1
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 840)
Protilátky
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 840
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human