Tlačiť
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 450)
Dodávateľ: AAT BIOQUEST
102800Z0 102800Z1
AATB102800Z0EA 397.11 EUR
AATB102800Z0 AATB102800Z1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 450)
Protilátky
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human