Tlačiť
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Red 780)
Dodávateľ: AAT Bioquest
100520W0 100520W1
AATB100520W0EA 397.11 EUR
AATB100520W0 AATB100520W1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Red 780)
Protilátky
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: mFluor™ Red 780
Epitope:
Host: Mouse
Isotype:
Reactivity: Human