Tlačiť

Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/iFluor® 647)

Dodávateľ: AAT BIOQUEST
104411Q0 104411Q1 104411Q2
AATB104411Q0EA 341.36 EUR
AATB104411Q0 AATB104411Q1 AATB104411Q2
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/iFluor® 647)
Protilátky
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Pozor: For research use only.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: PE (Phycoerythrin)/iFluor® 647
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR