Tlačiť

Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 540)

Dodávateľ: AAT BIOQUEST
10340120 10340121
AATB10340120EA 397.11 EUR
AATB10340120 AATB10340121
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 540)
Protilátky
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Pozor: For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR