Tlačiť
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Violet 450)
Dodávateľ: AAT BIOQUEST
101610Z0 101610Z1
AATB101610Z0EA 397.11 EUR
AATB101610Z0 AATB101610Z1
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (mFluor™ Violet 450)
Protilátky
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human