Tlačiť
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 750)
Dodávateľ: AAT BIOQUEST
100210L0 100210L1
AATB100210L0EA 397.11 EUR
AATB100210L0 AATB100210L1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 750)
Protilátky
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human