Tlačiť

Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 610)

Dodávateľ: AAT BIOQUEST
100210D0 100210D1
AATB100210D0EA 397.11 EUR
AATB100210D0 AATB100210D1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 610)
Protilátky
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Pozor: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 610
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR