Tlačiť
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 430)
Dodávateľ: AAT BIOQUEST
10032030 10032031
AATB10032030EA 397.11 EUR
AATB10032030 AATB10032031
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 430)
Protilátky
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 430
Epitope:
Host: Mouse
Isotype:
Reactivity: Human