Tlačiť
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 405)
Dodávateľ: AAT BIOQUEST
10161020 10161021
AATB10161020EA 397.11 EUR
AATB10161020 AATB10161021
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 405)
Protilátky
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Pozor: For research use only.
Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 405
Epitope:
Host: Mouse
Isotype:
Reactivity: Human