Tlačiť

Anti-Goat Polyclonal Antibody (CF® 583R)

Dodávateľ: BIOTIUM
20906-500UL 20906-50UL
20906-500ULEA 282.79 EUR
20906-500UL 20906-50UL
Anti-Goat Polyclonal Antibody (CF® 583R)
Protilátky
Highly cross-adsorbed goat anti-rat IgG (H L) secondary antibody labeled with our superior CF® dyes.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 16 bright and photostable CF® dyes
  • Biotin, AP, and HRP conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology

This is a highly cross-adsorbed goat anti-rat IgG (H L) secondary antibody labeled with our bright and photostable CF® tyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, horse, human, and rabbit serum.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: CF® 583R
Epitope:
Host: Goat
Isotype:
Reactivity: Rat IgG
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR