Tlačiť
GSK-3 inhibitor XIII 99 HPLC_ASSAY_METHOD
Dodávateľ: ENZO LIFE SCIENCES
ENZCHM1240001 ENZCHM1240005
ENZOENZCHM1240001EA 134.99 EUR
ENZOENZCHM1240001 ENZOENZCHM1240005
GSK-3 inhibitor XIII 99 HPLC_ASSAY_METHOD
GSK-3 inhibitor XIII
Potent inhibitor of GSK-3 ATP-binding site.
Mw: 301.4 g/mol
Číslo CAS: 404828-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=870f095cbcf6cd35145e536245534395f10ebf19&vid=db7c3376450f2c45c46394a2d481dc489d3e069f&prid=web&clid=SAPDAM