Tlačiť
DURAN® Reactor, Stirred, GLS 80 Thread
Dodávateľ: DWK LIFE SCIENCES
1200379 1200380 1200381 1200382 1200385 1200386 1200383 212639107
215-2468EA 423.99 EUR
215-2468 441-0550 441-0551 215-2473 215-2469 215-2470 215-2474 215-4158
DURAN® Reactor, Stirred, GLS 80 Thread
Baňky Baňky míchané
DURAN®, borosilicate glass 3.3, clear, PP, PTFE, PEEK, stainless steel.
  • Parts in contact with media conform fully to FDA requirements
  • The agitating element is exchangeable
  • With the aid of components from the DURAN® connection system extra media bottles can be connected or a sterile pressure release is possible
The GLS 80 stirred reactor is suitable for a wide range of laboratory mixing processes. The connections provided permit addition or removal of media from the bottle during the mixing process. The whole assembly can be autoclaved and is therefore suitable for use in biological applications. By using components from the GLS 80 connection system, an additional media bottle (OD hose: 1,6 to 12,0 mm) can be connected or a sterile pressure equaliser attached.
Drive for the stirrer is provided by a standard commercial magnetic stirrer. The variable agitator shaft can be used in DURAN® GLS 80 laboratory glass bottles (1000 and 2000 ml) and provides notably improved mixing in comparison with standard magnetic stirring bars. The agitating element is exchangeable and use up to 500 rpm is possible. Typical applications: Mixing of liquids, dissolving of solids, simple fermentation processes.