Tlačiť
Magnifier lamps, varioLED flex
Dodávateľ: ESCHENBACH OPTIK
27815 2781
630-2155EA 756.12 EUR
630-2155 630-2154
Magnifier lamps, varioLED flex
Skla zvětšovací /Lupy
These illuminated bench magnifiers are suitable for use in the scientific and professional fields, particularly for detailed technical work and reading.
  • Aspheric PXM® lightweight lens, cera-tec® coated for a distortion-free image with edge-to-edge sharpness
  • Segment switching provides a choice between ring illumination and segment illumination for closer inspection of different surface structures
  • Illumination level: approx. 6000 lx (at a working distance of 150 mm)
  • Gooseneck design with protective cover for improved hygiene
Low power consumption as power requirement is only 5 W, service life of LED approx. 50000 hours.