Tlačiť

Dimethylsulfoxid

Dodávateľ: SCHARLAB

Synonymá: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

SU01551000 SU01552500
SCARSU01551000EA 0 EUR
SCARSU01551000 SCARSU01552500
Dimethylsulfoxid
Dimethylsulfoxid
Vzorec: (CH₃)₂SO
Mw: 78.14
Teplota varu: 189
Teplota tavenia: 18.5
Hustota: 1.101 g/cm³ (20 °C)
Teplota vzplanutia: 87
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002089
Číslo CAS: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR