Tlačiť

N,N-Diethylmethylamine, Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ200

D98203-100G D98203-25G
SIALD98203-100GEA 90.65 EUR
SIALD98203-100G SIALD98203-25G
N,N-Diethylmethylamine, Sigma-Aldrich®
N,N-Diethylmethylamine
Vzorec: C₅H₁₃N
Teplota varu: 63 °C (1013 hPa)
Teplota tavenia: –196 °C
Hustota: 0,7227 g/cm³ (20 °C)
Číslo MDL: MFCD00009049
Číslo CAS: 616-39-7
EINECS: 210-480-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR