Tlačiť

N-Methylsuccinimide, Sigma-Aldrich®

Dodávateľ: Merck

Synonymá: 1-Methylpyrrolidine-2,5-dione

M-Clarity™ quality level = MQ100

325384-5G 325384-25G
SIAL325384-5GEA 90.65 EUR
SIAL325384-5G SIAL325384-25G
N-Methylsuccinimide, Sigma-Aldrich®
N-Methylsuccinimide
Vzorec: C₅H₇NO₂
Teplota varu: 234 °C (1013 hPa)
Teplota tavenia: 66…69 °C
Hustota: 1,37 g/cm³ (20.85 °C)
Číslo MDL: MFCD00005517
Číslo CAS: 1121-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR