Dysprosium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
Synonymá:
Dysprosium (III) nitrate pentahydrate
11315.14
11315.22
11315.14EA
217.81
EUR
11315.14
11315.22
Dysprosium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dysprosium trinitrate pentahydrate
Purity: 99.99%
CAS No.: 10031-49-9
Molecular Formula: Dy(NO3)3.5H2O
Formula Weight: 438.59 (348.51anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Cream to pale yellow
Odour: Odourless
pH: 3
Melting Point: 88.6°
Merck Reference: 14,3482
UN No.: 1477
MDL No.: MFCD00149683
Vzorec:
DyN₃O₉·5H₂O Mw: 438.59 Teplota tavenia: 88.6 °C Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00149683 Číslo CAS: 10031-49-9 EINECS: 233-410-5 OSN: 1477 ADR: 5.1 III Merck Index: 14,3482 |