Screen Quest™ Rhod-4 No Wash Calcium Assay Kits, Medium Removal
Dodávateľ: AAT BIOQUEST
36330
36331
36332
AATB36330EA
162.86
EUR
AATB36330
AATB36331
AATB36332
Screen Quest™ Rhod-4 No Wash Calcium Assay Kits, Medium Removal
Test
Buněčná stanovení
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- >250 times fluorescence increase once bound to calcium
- Long-wavelength signal emission wavelength
- Contains all necessary components
- No wash step required
- Robust and adaptable protocol
- brighter and more photostable signal
Pozor: For research use only.