Tlačiť
ReadiUse™ Rapid Luminometric ATP Assay Kits
Dodávateľ: AAT Bioquest
21601 21602 21603
AATB21601EA 162.86 EUR
AATB21601 AATB21602 AATB21603
ReadiUse™ Rapid Luminometric ATP Assay Kits
Test Buněčná stanovení
Adenosine triphosphate (ATP) plays a fundamental role in cellular energetics, metabolic regulation and cellular signaling.
  • Extreme sensitivity
  • Stable signal with >2 hour half-life
  • No DTT used
Pozor: For research use only.