Tlačiť

Goat Anti-Rat IgG2b Secondary Antibody

Dodávateľ: Novus Biologicals
NOVUNBP2-68490EA 0 EUR
NOVUNBP2-68490
Goat Anti-Rat IgG2b Secondary Antibody
Protilátky
Goat anti Rat IgG2b antibody recognises rat IgG2b as assessed by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG2b from other species.

Recommended Dilutions: ELISA: 1 to 10 µg/ml, Flow Cytometry, Western Blot
Preservative: 0.09% Sodium Azide

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR