Tlačiť

Anti-HNF-3 alpha/FoxA1 Human Recombinant Antibody

Dodávateľ: Nova Biomedical
NOVUNBP2-38628PEPEA 426.53 EUR
NOVUNBP2-38628PEP
Anti-HNF-3 alpha/FoxA1 Human Recombinant Antibody
Protilátky
Antigen: HNF-3 alpha/FoxA1
Clonality: Recombinent
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR