Tlačiť
Thermometer Temperature Indicating Label
Dodávateľ: BRADY
195837 195838
BRDY195837EA 47.67 EUR
BRDY195837 BRDY195838
Thermometer Temperature Indicating Label
Proužky indikátorů teploty
Temperature indicating labels are a cost-effective solution for temperature checks during maintenance or for trouble spotting during research and development. The labels extremely low profile allows for application on components, on industrial and household applications such as electric pumps, rotating equipment, brakes and refrigerators, in cold chains and hot spaces or in areas where measuring temperature becomes difficult or impractical.
  • Sub-surface printed polyester
  • Gloss finish
  • Cost effective
Application like cold temperature labelling, componentMore Product Information