Tlačiť
Erlenmeyer Flasks, Narrow Neck
Dodávateľ: GLASSCO
231.202.01 231.202.02 231.202.05 231.202.06 231.202.07
GLAS231.202.01EA 2.15 EUR
GLAS231.202.01 GLAS231.202.02 GLAS231.202.05 GLAS231.202.06 GLAS231.202.07
Erlenmeyer Flasks, Narrow Neck
Baňky Baňky dle Erlenmeyera
Clear, conical, graduated and narrow mouth Erlenmeyer flask, made from ASTM E-438 Type 1, Borosilicate 3.3 heat resistance glass.
  • All the flasks are printed in white enamel
  • Has reinforced thick rim to reduce chipping and breakage
Certifikácie: Complies with ISO 1773.