Tlačiť
Heat Resistant Polyimide Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Dodávateľ: BRADY
174188 173777 173714 174178 173885 174254 174204 173866 173897
BRDY174188EA 320.1 EUR
BRDY174188 BRDY173777 BRDY173714 BRDY174178 BRDY173885 BRDY174254 BRDY174204 BRDY173866 BRDY173897
Heat Resistant Polyimide Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Štítky
High temperature polyimide (B-727) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Ultra-durable adhesive is designed for use with extreme wash protocol and cleaning chemicals
  • Ideal for use in auto apply equipment
  • Glossy white polyimide material is thermal transfer printable
  • Service temperature of −70 to +100 °C (−94 to +212 °F)
Ideal for circuit board labelling, component and equipment labelling, and temperature indicating labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 Label Printers or M7-R6000 for the M710/BMP71 Label Printers. This label is no longer compatible with TLS series printers.
Certifikácie: UL recognised. Meets the performance requirements of MIL -STD-202G, Notice 12, Method 215K. Dibutyltin- and Dioctyltin-free.