Tlačiť

Anti-Factor VIII Rabbit Polyclonal Antibody

Dodávateľ: Novus Biologicals
NOVUNB100-91761EA 701.35 EUR
NOVUNB100-91761
Anti-Factor VIII Rabbit Polyclonal Antibody
Protilátky
The Factor VIII Antibody from Novus Biologicals is a rabbit polyclonal antibody to Factor VIII. This antibody reacts with human, mouse, rat. The Factor VIII Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 to 1:200

Type: Primary
Antigen: Factor VIII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR