Tlačiť
Cleanroom gloves, polychloroprene, sterile, Biogel® NeoTech Indicator® underglove
Dodávateľ: MOLNLYCKE HEALTH CARE
4440655 4440660 4440665 4440670 4440675 4440680 4440685 4440690
112-6346EA 583.23 EUR
112-6346 112-6347 112-6348 112-6349 112-6350 112-6351 112-6352 112-6353
Cleanroom gloves, polychloroprene, sterile, Biogel® NeoTech Indicator® underglove
Rukavice
These sterile, blue cleanroom undergloves with beaded cuffs and curved fingers are made to be worn in combination with the Biogel® NeoTech overglove to form a Breach Indication System. The gloves are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments.
  • Exceed industry standard for force at break, tensile strength and elongation
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polychloroprene
  • Biogel® hydrogel polymer coating for easy donning
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certifikácie: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Balenie: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.