Tlačiť
Silicone Keyboards with Integrated Touchpad, InduProof® Advanced TOUCH
Dodávateľ: GETT GERATETECHNIK
KG18216 KG19221
GETTKG18216EA 335.34 EUR
GETTKG18216 GETTKG19221
Silicone Keyboards with Integrated Touchpad, InduProof® Advanced TOUCH
Klávesnice
Rugged, waterproof and dustproof silicone keyboard with integrated touchpad.
  • Rugged, durable keyboard with 104 keys and integrated touchpad
  • Comfortable and ergonomic operation, even with protective gloves (silicone or latex)
  • IP 68 protection; device can be submerged into liquids
  • Easy cleaning and disinfection, even under tap water or by immersion
  • Increased chemical and mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75mm)
  • 4-year warranty on technical function
  • Made in Germany
The InduProof® Advanced keyboard has a complete set of keys with numeric keypad and an integrated touchpad. It provides a pleasant tactile feel and the highest degree of protection. The robust design makes it ideal for use in demanding hygiene sensitive areas such as industrial food and beverage production. High-quality workmanship and the use of high-quality materials in this keyboard ensure a long service life.
Certifikácie: CE, REACH, RoHs, EMC, IP 68.
Balenie: Keyboards are safely and environmentally friendly packed in brown cardboard boxes.