Tlačiť
Precision (milligram) laboratory balances, Cubis® II MCE
Dodávateľ: Sartorius Balances
MCE10202S2S000 MCE1203S2S00-A MCE1202S2S00-0 MCE623S2S00-R MCE5203S2S00-E MCE5203P2S00-E MCE3203S2S00-E MCE2203S2S00-E MCE2203P2S00-E MCE1203S2S00-E MCE623S2S00-E MCE623P2S00-E MCE323S2S00-E MCE5203S2S00-R MCE5203P2S00-R MCE3203S2S00-R MCE2203S2S00-R MCE2203P2S00-R MCE1203S2S00-R MCE623P2S00-R MCE323S2S00-R MCE5202S2S00-E MCE5202S2S00-R MCE14202S2S000 MCE14202P2S000 MCE12201S2S000 MCE11201S2S000 MCE5203S2S00-A MCE5203P2S00-A MCE3203S2S00-A MCE2203S2S00-A MCE2203P2S00-A MCE623S2S00-A MCE623P2S00-A MCE323S2S00-A MCE5203S2S00-I MCE5203P2S00-I MCE3203S2S00-I MCE2203S2S00-I MCE2203P2S00-I MCE1203S2S00-I MCE623S2S00-I MCE623P2S00-I MCE323S2S00-I MCE5203S2S00-U MCE5203P2S00-U MCE3203S2S00-U MCE2203S2S00-U MCE2203P2S00-U MCE1203S2S00-U MCE623S2S00-U MCE623P2S00-U MCE323S2S00-U MCE6202S2S00-0 MCE6202P2S00-0 MCE4202S2S00-0 MCE8201S2S00-0 MCE5202S2S00-A MCE5202S2S00-I MCE5202S2S00-U MCE14202S-2CEU MCE14202S-2S00 MCE12201S2CFR0
SARBMCE10202S2S000EA 10538.31 EUR
SARBMCE10202S2S000 SARBMCE1203S2S00-A SARBMCE1202S2S00-0 SARBMCE623S2S00-R SARBMCE5203S2S00-E SARBMCE5203P2S00-E SARBMCE3203S2S00-E SARBMCE2203S2S00-E SARBMCE2203P2S00-E SARBMCE1203S2S00-E SARBMCE623S2S00-E SARBMCE623P2S00-E SARBMCE323S2S00-E SARBMCE5203S2S00-R SARBMCE5203P2S00-R SARBMCE3203S2S00-R SARBMCE2203S2S00-R SARBMCE2203P2S00-R SARBMCE1203S2S00-R SARBMCE623P2S00-R SARBMCE323S2S00-R SARBMCE5202S2S00-E SARBMCE5202S2S00-R SARBMCE14202S2S000 SARBMCE14202P2S000 SARBMCE12201S2S000 SARBMCE11201S2S000 SARBMCE5203S2S00-A SARBMCE5203P2S00-A SARBMCE3203S2S00-A SARBMCE2203S2S00-A SARBMCE2203P2S00-A SARBMCE623S2S00-A SARBMCE623P2S00-A SARBMCE323S2S00-A SARBMCE5203S2S00-I SARBMCE5203P2S00-I SARBMCE3203S2S00-I SARBMCE2203S2S00-I SARBMCE2203P2S00-I SARBMCE1203S2S00-I SARBMCE623S2S00-I SARBMCE623P2S00-I SARBMCE323S2S00-I SARBMCE5203S2S00-U SARBMCE5203P2S00-U SARBMCE3203S2S00-U SARBMCE2203S2S00-U SARBMCE2203P2S00-U SARBMCE1203S2S00-U SARBMCE623S2S00-U SARBMCE623P2S00-U SARBMCE323S2S00-U SARBMCE6202S2S00-0 SARBMCE6202P2S00-0 SARBMCE4202S2S00-0 SARBMCE8201S2S00-0 SARBMCE5202S2S00-A SARBMCE5202S2S00-I SARBMCE5202S2S00-U 611-5267 611-5268 SARBMCE12201S2CFR0
Precision (milligram) laboratory balances, Cubis® II MCE
Váhy
Cubis® II MCE precision balances with essential user interface offer a maximum load between 320 and 14200 g and a readability of 1 to 100 mg provide, ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis® II balance series from Sartorius.
All weighing modules of the Cubis® II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
These MCE models with essential user interface have a black and white touch screen plus holders for ergonomic sample handling and automatic motorised levelling. The balances feature innovative draft shield technology, gesture sensor and climate monitoring. With complete traceability the balances help achieve 21 CFR Part 11 and EU Annex 11 compliance.
Choice of draft shield denoted by model name suffix: manual glass doors with large draft shield (U), manual glass doors with standard draft shield (E), stainless steel draft shield (R), motorised draft shield (A) or motorised with built-in de-ioniser (I) for effective removal of static charges.