Tlačiť

Rabbit Polyclonal anti-Pichia Pastoris antibody

Dodávateľ: Novus Biologicals
NOVUNB100-65067EA 662.83 EUR
NOVUNB100-65067
Rabbit Polyclonal anti-Pichia Pastoris antibody
Protilátky
The Pichia Pastoris Antibody from Novus Biologicals is a rabbit polyclonal antibody to Pichia Pastoris. This antibody reacts with yeast. The Pichia Pastoris Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Pichia Pastoris
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR