Tlačiť

Mouse monoclonal anti-NFAT1 antibody (25A10.D6.D2)

Dodávateľ: Novus Biologicals
NOVUNB300-504EA 694.6 EUR
NOVUNB300-504
Mouse monoclonal anti-NFAT1 antibody (25A10.D6.D2)
Protilátky
The NFATC2 / NFAT1 Antibody (25A10.D6.D2) from Novus Biologicals is a mouse monoclonal antibody to NFATC2 / NFAT1. This antibody reacts with human, mouse, rat. The NFATC2 / NFAT1 Antibody (25A10.D6.D2) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Sandwich ELISA.

Type: Primary
Antigen: NFATC2/NFAT1
Clonality: Monoclonal
Clone: 25A10.D6.D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR