Tlačiť
Powered air respirator head, neck, and shoulder covers, Versaflo™
Dodávateľ: 3M
Versaflo™
MMMAM972EA 71.09 EUR
MMMAM972
Powered air respirator head, neck, and shoulder covers, Versaflo™
Polomasky a filtrační respirátory Systémy polomasek
The Versaflo™ Headtop Cover M-972 is a flame-resistant polyester headtop cover designed to protect the M-Series headtop from sparks and hot particles.
  • Flame-resistant headtop cover
  • Use with any Versaflo™ M-Series product
Protect the outside surface of the helmet shell from sparks and hot particles with the Versaflo™ Flame Resistant Headtop Cover, M-972. This cover is made from flame-resistant polyester and is a disposable item. It should be discarded when it becomes damaged or soiled.