Tlačiť
Masterflex® Flange Kits for Mechanically Coupled Centrifugal Pumps, Avantor®
Dodávateľ: Avantor Fluid Handling
70725-70 70725-72
MFLX70725-70EA 595.67 EUR
MFLX70725-70 MFLX70725-72
Masterflex® Flange Kits for Mechanically Coupled Centrifugal Pumps, Avantor®
Čerpadla Odstředivá čerpadla
Enhance the life of your pump with service flange kits.
  • Constructed of stainless steel (SS) parts
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.
Informace o dodávce: Includes stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.