Tlačiť
Parker Hannifin Reducer Fittings, Compression, Straight
Dodávateľ: Parker Hannifin
MFLX30486-10EA 63.54 EUR
MFLX30486-10 MFLX31320-05 MFLX30486-13 MFLX31320-03 MFLX31320-04 MFLX31320-01 MFLX31320-02 MFLX31320-00 MFLX30486-08 MFLX30486-15
Parker Hannifin Reducer Fittings, Compression, Straight
Fitinky a konektory
Get a secure seal at higher pressures.
  • Quick and easy assembly without tools
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.