Tlačiť
Masterflex® L/S® 2-Stop Pump Tubing, FDA-Compliant Viton®, Avantor®
Dodávateľ: Avantor Fluid Handling
MFLX96428-13EA 194.27 EUR
MFLX96428-13 MFLX96428-15 MFLX96428-16 MFLX96428-24 MFLX96428-25 MFLX96428-35
Masterflex® L/S® 2-Stop Pump Tubing, FDA-Compliant Viton®, Avantor®
Hadičky Hadičky peristaltické pumpy
Good for food and lab applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
Characteristics: Good chemical resistance, opaque black
For use with Masterflex® multichannel pump heads.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.