Tlačiť
ESD ankle boots, MARTEN XXSports Pro BOA®, mid, S3
Dodávateľ: ELTEN
768141/35 768141/36 768141/37 768141/38 768141/39 768141/40 768141/41 768141/42 768141/43 768141/44 768141/45 768141/46 768141/47 768141/48
ELTN768141/35EA 144 EUR
ELTN768141/35 ELTN768141/36 ELTN768141/37 ELTN768141/38 ELTN768141/39 ELTN768141/40 ELTN768141/41 ELTN768141/42 ELTN768141/43 ELTN768141/44 ELTN768141/45 ELTN768141/46 ELTN768141/47 ELTN768141/48
ESD ankle boots, MARTEN XXSports Pro BOA®, mid, S3
Obuv
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole SPORTIVE ESD black
  • Non-metallic, penetration-resistant midsole
  • TPU/PU sole WELLMAXX SPORTS
  • TPU scuff cap
  • Steel toe cap
  • BOA® Fit System
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.