Tlačiť
ESD safety shoes, MADDOX, low, S3
Dodávateľ: ELTEN
729391/35 729391/36 729391/37 729391/38 729391/39 729391/40 729391/41 729391/42 729391/43 729391/44 729391/45 729391/46 729391/47 729391/48
ELTN729391/35EA 114.32 EUR
ELTN729391/35 ELTN729391/36 ELTN729391/37 ELTN729391/38 ELTN729391/39 ELTN729391/40 ELTN729391/41 ELTN729391/42 ELTN729391/43 ELTN729391/44 ELTN729391/45 ELTN729391/46 ELTN729391/47 ELTN729391/48
ESD safety shoes, MADDOX, low, S3
Obuv
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD grey
  • Non-metallic, penetration-resistant midsole
  • TPU/PU sole MAXXIMO
  • Steel toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.