Tlačiť
ESD safety shoes, lace-up, BRANDON XXTM Low
Dodávateľ: ELTEN
720511/36 720511/37 720511/38 720511/39 720511/40 720511/41 720511/42 720511/43 720511/44 720511/45 720511/46 720511/47 720511/48
ELTN720511/36EA 108.3 EUR
ELTN720511/36 ELTN720511/37 ELTN720511/38 ELTN720511/39 ELTN720511/40 ELTN720511/41 ELTN720511/42 ELTN720511/43 ELTN720511/44 ELTN720511/45 ELTN720511/46 ELTN720511/47 ELTN720511/48
ESD safety shoes, lace-up, BRANDON XXTM Low
Obuv
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised microfibre
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO grey
  • Non-metallic, penetration-resistant midsole
  • MONO-PU sole WELLMAXX TRAINERS MONO
  • Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.
Certifikácie: EN ISO 20345 S3 SRC, form A