Tlačiť
Culture media, ready to use, Compact Dry
Dodávateľ: Shimadzu Diagnostics Europe
54061-0AQ-0040 54061-0AQ-0240 54061-0AQ-1400 54053-0CF-0040 54053-0CF-0240 54053-0CF-1400 54052-0EC-0040 54052-0EC-0240 54052-0EC-1400 54055-ETB-0040 54055-ETB-0240 54055-ETB-1400 54056-ETC-0040 54056-ETC-0240 54056-ETC-1400 54060-0LS-0040 54060-0LS-0240 54060-0LS-1400 54062-0PA-0040 54062-0PA-0240 54062-0PA-1400 54058-0SL-0040 54058-0SL-0240 54058-0SL-1400 54051-0TC-0040 54051-0TC-0240 54051-0TC-1400 06749-0VP-0040 06749-0VP-0240 54068-0BC-0040 54068-0BC-0240 54068-0BC-1400 54057-XSA-0040 54057-XSA-0240 54057-XSA-1400 54054-0YM-0040 54054-0YM-0240 54063-YMR-0040 54063-YMR-0240 54063-YMR-1400
710-2189EA 147.61 EUR
710-2189 710-2190 710-2191 710-2168 710-2169 710-2170 710-2165 710-2166 710-2167 710-2174 710-2175 710-2176 710-2177 710-2178 710-2179 710-2186 710-2187 710-2188 710-2192 710-2193 710-2194 710-2183 710-2184 710-2185 710-2162 710-2163 710-2164 710-2160 710-2161 710-2204 710-2205 710-2206 710-2180 710-2181 710-2182 710-2171 710-2172 710-2195 710-2196 710-2197
Culture media, ready to use, Compact Dry
Média mikrobiologická Připravená mikrobiologická média
Ready to use dehydrated film media in rigid plastic with lid for cultivating and counting microorganisms in food, for monitoring product quality and potential contamination. These plates can be used to test raw material as well as finished products. Microorganisms are identified by the colours that are developed by chromogenic substrates and redox indicators.
  • Ready to use - place 1 ml sample onto the plate and incubate
  • Easy to read - colonies are pigmented with different colours
  • Easy to store at room temperature - long shelf life
  • Space saving and stackable
Dry TC, EC, CF,YM, XSA, SL, LS are AOAC approved
Dry TC, EC, CF, ETB, XSA, SL, LS are MicroVal approved
Dry TC, EC, CF, ETB, XSA, SL, LS are NordVal approved
European validation and certification alternative methods acc. to EN ISO 16140