Tlačiť
Full face masks with Typhoon™ valve, Force®10
Dodávateľ: JSP
BPB003-004-000 BPB003-104-000 BPB003-204-000
111-2600EA 296.03 EUR
111-2600 111-2601 111-2602
Full face masks with Typhoon™ valve, Force®10
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Masky celoobličejové
The Force®10 Typhoon™ is an extremely lightweight, easy to maintain, full face mask with panoramic visor. With 3 available sizes (S,M,L) and the ability to fit PressToCheck™ filters, the user can feel reassured of an excellent fit.
  • Available in Small, Medium, or Large sizes for a perfect fit
  • Compatible with PressToCheck™ Filters (not included)
  • Typhoon™ exhalation valve for reduced breathing resistance
  • Polycarbonate lens, ABS buckles
  • Engineered for easy maintenance
  • One of the lightest full face masks on the market
The Force®10 Typhoon™ has been meticulously engineered to incorporate a small number of key components. Disassembly, cleaning, and maintenance are effortless with replacement parts available if necessary. Compatible with both JSP Classic filters and the PressToCheck™ filters, which allow for daily face fit reassurance.
Comfort is a key feature of this ultra-lightweight mask, as the web-like structure of the harness cradles the users head, ensuring wearer comfort throughout the working day. The addition of different sizing options allows for a more tailored fit and greater comfort.
The unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures (−40 °C tested).
Certifikácie: Facepiece EN136. Gas Filter Performance EN14387. Particulate Filter Performance EN143. Flammability EN136.