Tlačiť
ESD safety shoes, lace-up, uvex 1 G2 planet, 6823
Dodávateľ: Uvex
uvex 1 G2 planet
114-9109EA 161.79 EUR
114-9109 114-9108 114-9119 114-9107 114-9118 114-9113 114-9112 114-9111 114-9121 114-9110 114-9106 114-9117 114-9116 114-9105 114-9104 114-9115 114-9114 114-9120
ESD safety shoes, lace-up, uvex 1 G2 planet, 6823
Obuv
These extremely lightweight S1 P safety shoes are made from recycled and sustainable materials.
  • Made from synthetic materials, making it suitable for people allergic to chrome
  • Meets the ESD specification with volume resistance under 35 megaohms
  • Elastic laces for quick, individual adjustment (normal laces also provided)
  • Soft padding on dust tongue and collar
  • Virtually seam-free upper design made from textile material to prevent pressure points
  • uvex x-dry knit planet textile material made from 100% recycled PET bottles
  • New uvex xenova® composite toe cap and metal-free, penetration-resistant midsole, does not restrict the flexibility of the shoe
The new, innovative uvex i-PUREnrj sole technology offers the highest level of shock absorbing properties at the forefoot and at the heel, resulting in excellent rebound over the entire midsole and optimal stability thanks to the foam heel basket. A new, wider, 100% metal-free uvex xenova® safety toe cap provides more toe room and an optimum fit. This toe cap offers compact design, anatomical shape, good lateral stability, and no thermal conductivity. All sole materials are free of silicone, plasticisers, and other paint wetting impairment substances.
uvex x-tended grip planet TPU outsole made from up to 10% reprocessed PU waste from our production, uses the latest in biomechanical research, is extremely durable and, therefore, very slip resistant, with tread that makes it particularly suitable for use on industrial floors. Non metallic, penetration resistant midsole made from 100% recycled material, in accordance with the latest standards, does not restrict the flexibility of the shoe.
Removable anti-static comfortable insole with a surface made from 100% recycled PET bottles and foam made from reprocessed PU production waste and recycled PU, with a moisture transport system and additional heel and forefoot shock absorption as well as good arch support. TPU scuff cap made from 40% recycled material protects the outer shoe material from wear. Breathable lining made from 18% bamboo and 52% recycled polyester for optimised climate management.
Width: 11
Certifikácie: EN ISO 20345:2011 S1 P SRC.